Learn More
Electron Microscopy Sciences Serum Tube Vials Clear, 20 ml
Supplier: Electron Microscopy Sciences 6030020
Wheaton glass containers are the choice for lyophilization. They excel in the three majors factors that contribute to successful Lyophillization: heat transfer, sealability, & water vapor transmission barrier. Heat Transfer–a Wheaton Glass serum vial, as opposed to glass serum bottles, plastic serum bottles, & other containers has a very consistent thin wall & bottom that allows the heating phase of the lyophilization process to take place rapidly & effectively. Sealability–Wheaton glass serum vials feature a inblowbackin feature in neck & bottom that allows for a 2 or 3 legged lyophilization stopper to be securely & evenly seated in the mouth of the vial. Water vapor Transmission Barrier–Wheaton glass serum vials are made from premium, low extractable, borosilicate glass, which offers a superiorbarrier to water vapors. These vials can be autoclaved, by dry heat, by chemical, by gas, or by radiation.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.