Learn More
Enzo Life Sciences SCREEN-WELL Serotonergic ligand library (100µl/well)
Supplier: Enzo Life Sciences BML28130100
The Serotonergic Ligands Plate contains 79 ligands including endogenous neurotransmitters, agonists, antagonists, and marketed drugs. The library is ideal for screening or identifying recombinant orphan G protein coupled receptors, target validation, secondary screening, assay development, and other pharmacological applications. The Serotonergic Ligands plate is 1 plate of the larger Neurotransmitter Library (BML-2810) which contains over 680 ligands. Long Term Storage: -80°C. Applications: HTS .
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.