Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ SB 224289 hydrochloride
Selective 5-HT1B antagonist
$306.24 - $1289.92
Specifications
Chemical Name or Material | 1'-Methyl-5-[[2'-methyl-4'-(5-methyl-1,2,4-oxadiazol-3-yl)biphenyl-4-yl]carbonyl]-2,3,6,7-tetrahydrospiro[furo[2,3-f]indole-3,4'-piperidine hydrochloride |
---|---|
CAS | 180084-26-8 |
Assay Percent Range | >95% |
Solubility | Soluble to 10mM with gentle warming in DMSO |
Molecular Formula | C32H32N4O3.HCl |
Description
Selective 5-HT1B receptor antagonist (pKi = 8.2). Displays > 60-fold selectivity over 5-HT1D, 5-HT1A, 5-HT1E, 5-HT1F, 5-HT2A and 5-HT2C receptors in radioligand binding and functional assays. Centrally active following oral administration in vivo.Specifications
1'-Methyl-5-[[2'-methyl-4'-(5-methyl-1,2,4-oxadiazol-3-yl)biphenyl-4-yl]carbonyl]-2,3,6,7-tetrahydrospiro[furo[2,3-f]indole-3,4'-piperidine hydrochloride | |
>95% | |
C32H32N4O3.HCl |
180084-26-8 | |
Soluble to 10mM with gentle warming in DMSO | |
Observed MW: 557.09 |
Safety and Handling
Recommended Storage : Store at -20°C
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title