Learn More
Biocare Medical, LLC Rat-on-Mouse HRP (horseradish peroxidase)-Polymer, 6ml
Supplier: Biocare Medical, LLC RT517G
The Rat-on-Mouse HRP-Polymer is for use with rat primary antibodies on mouse tissues. This detection system is mouse adsorbed for minimum cross-reactivity to endogenous mouse IgG. It is labeled with horseradish peroxidase (HRP).