Learn More
Hettich Centrifuges ROTIXA500,(120)13(120)16,BioPk
Supplier: Hettich Centrifuges ROTIXABLOOD4BC
Hettich ROTIXA 500 RS BLOOD TUBE PACKAGE 4 BIO-CONTAINMENT (1) Centrifuge, (1) 90° Swing-out Rotor, (4) Buckets, (4) Adapters 13's & 16's, (4) Bio-containment Lids, Power: 220 V 1~60Hz, Warranty: Five (5) years