Learn More
Bioworld Rose Modified Multiplication Basal Medium, 25 L
Supplier:  Bioworld 306301743
Encompass_Preferred
Rose Modified Multiplication Basal MediumStage IIContains Micronutrients and Macronutrients required for rose multiplicationContains FeNaEDTA in place of Ferrous Sulfate and Disodium EDTASucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 4.51 g/LpH: 3.0–4.0Soluble in water
Catalog No. 50-254-932