Learn More
Roosterbio Inc Xeno-Free RoosterVial™-hAD: Xeno-Free Human Adipose-Derived Mesenchymal Stem/Stromal Cells
SDP
Supplier:  Roosterbio Inc C46010AD
Encompass_Preferred
RoosterVial™-hAD-XF are Human Adipose-Derived Mesenchymal Stem/Stromal Cells (hAD-MSCs) isolated from adult human adipose from a variety of normal, healthy donors and expanded to a population doubling level (PDL) of 6-10, post-mononuclear cell isolation.  Available in both 1M or high-volume 10M cell per vial formats to meet your needs throughout product development. Each lot is release tested to expand >10-fold within 4 to 6 days in a media exchange free process when paired with RoosterNourish™-MSC-XF bioprocess medium and conforms to ISCT minimal criteria for hMSC identity. Accompanying QC briefs gives added confidence in quality, reproducibility, and ease of integration. Designed for robust, reproducible, standardized scale-up and supported by our media exchange free 2D and 3D bioreactor processes and RoosterNourish™-MSC-XF expansion media, RoosterVial™-hAD-XF can efficiently and reproducibly get you the millions or billions of hMSCs to accelerate
Catalog No. NC2589088