Learn More
Medchemexpress LLC HY-116009 10mg Medchemexpress, RMC-4550 CAS:2172651-73-7 Purity:>98%
Supplier: Medchemexpress LLC HY11600910MG
Medchemexpress, HY-116009 10mg RMC-4550 CAS:2172651-73-7 RMC-4550 is a potent, selective and allosteric inhibitor of SHP2, with an IC50 of 0.583 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.