Learn More
World Precision Instrument Reusable Rapid Punch Biopsy Kit with Replacement
Supplier: World Precision Instrument WP1010
The Rapid-Punch consists of a razor sharp cutting tip designed to cut, retrieve and store cored samples from source materials such as tissue, gels, paper, cloth, leaves, paint chips, films or other thin substrates. The Rapid-Punch is recommended as the special DNA sampling tool. It is ideal for forensic and most laboratory sampling purposes. Designed for heavy, numerous and precise sampling operations. It is durable, lasting up to 2000 punches.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.