Learn More
MISCO REFRACTOMETER REFRACTOMETER CALIBRATN FLUID
Supplier: MISCO REFRACTOMETER CALFLUPA
Refractometer Calibration Fluid for MISCO Digital Handheld Refractometer-Special refractometer calibration fluid intended for calibrating the span (high-end) point on MISCO Palm Abbe digital refractometers.
This item is not returnable.
View return policy