Learn More
Chemglass Life Sciences 75L Process Reactor, Complete, Electric Motor, 90.8" OAH with Motor, 18.4" Clearance Below Valve, Supplied Complete With Manifold and Shielding
Supplier: Chemglass Life Sciences CG1968675
75L Process Reactor, Complete, Electric Motor, 90.8" OAH with Motor, 18.4" Clearance Below Valve, Supplied Complete With Manifold and Shielding. Complete reactor system on Chemglass's innovative Kwik Mount support frame. Lexan safety shields surround the reactor with the front panel hinged for easy access. Features: Rugged 1" Stainless Steel Pipe (1.315" OD) Uprights and Heavy Duty PTFE Coated Aluminum Fittings Provide Maximum Strength and Chemical Durability. Digital Temperature Monitor. Electric Motor has Separate Controller with Digital Display. Reactor Jacket Graduation Scale on Both Sides. Zero Dead Space Detachable Drain Valve has Knob With Laser Etched Directional Arrows for Open And Close. 7-Neck Lid with Large Powder Addition Port. Reactor Support Plate Holds Reactor Securely and Allows Easy Installation/Removal of Reactor. Lexan Safety Shield on All Four Sides. Supplied With Insulated Hose and Bulkhead Fittings.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.