Learn More
Creative Biomart R-Spondin-1 (Rspo-1) Protein
Supplier: Creative Biomart THP0281
Recombinant Human R-Spondin-1 is a 26.7 kDa protein consisting of 243 amino acid residues. Due to glycosylation, R-Spondin-1 migrates at an apparent molecular weight of approximately 40.0 kDa by SDS-PAGE analysis under reducing conditions.