Learn More
Perkin Elmer US LLC Un-Assembled Aluminum Crimp Cap with PTFE/Butyl (Red) Septa and Star Spring, 100/pk
Supplier: Perkin Elmer US LLC B0104239
For sealing sample vials for temperatures up to 130°C. Very low background peaks, lowest permeability, not recommended for repetitive injections from the same vial or for storage.