Learn More
Sigma Aldrich Fine Chemicals Biosciences Toxoplasma gondii Profilin | Purity: >95% | Mol Wt: 14.5 kDa | CAS No: 120224-57-0 | MDL No: MFCD01329910 | 1 MG
Supplier: Sigma Aldrich Fine Chemicals Biosciences SRP805010UG
Toxoplasma gondii profilin | Purity: >95% | Mol Wt: 14.5 kDa | CAS No: 120224-57-0 | MDL No: MFCD01329910 | 1 mg