Learn More
Perkin Elmer US LLC Preassembled Semi-Demountable Cell Kit - KBr, 0.5 mm
Supplier: Perkin Elmer US LLC L1270758
Preassembled Demountable Cells have rectangular windows, which allows these cells to function as either a semi-demountable cell or as a sealed liquid cell. Each cell has a set path length and cell window material. These cells are compatible with Spectrum 65, Spectrum 400, Spectrum One, Spectrum 100 spectrometers.