Learn More
Waters Corp Polystyrene Standard 20,000,000
Supplier: Waters Corp WAT011630
An individual GPC standard (Gel Permeation Chromatography standards) for molecular weight determination. Polystyrene calibration standards for use in characterizing or confirming your chemical analysis. This high-quality, proven standards provide confidence in your results. Polystyrene at 20,000,000 Da