Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Polymedco™ Poly stat™ Strep A Test
Lateral flow, rapid one-step test for qualitative detection of Group A Streptococcal antigen
Specifications
Clia Complexity | Waived |
---|---|
Detectable Analytes | Streptococcal Antigen (Group A) |
Sample Type | Throat Swab |
Quantity | 25 Pack |
Alert:
Contains sodium azide which may cause irritation to skin and eyes, mucous membranes and upper respiratory tract.
Catalog Number | Mfr. No. | Format | Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Format | Type | Price | Quantity & Availability | ||||
21-000-129
|
Polymedco™
STS25 |
Strip | Poly Stat Streptococcus A Test Strip |
Pack of 25 for $233.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
21-000-130
|
Polymedco™
STF25 |
Flip | Poly Stat Streptococcus A Test Flip |
Pack of 25 for $233.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
- Direct detection from throat swabs
- Simple visual result
- Available in flip and strip formats
Certifications
CLIA Complexity: Waived (Flip format), For waived facilities (Strip format)
Specifications
Waived | |
Streptococcal Antigen (Group A) | |
Throat Swab | |
25 Pack |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title