Learn More
Waters Corp Polyethylene Oxide Standard 79,000
Supplier: Waters Corp WAT011578
An individual GPC standards (Gel Permeation Chromatography standards) for molecular weight determination. Polyethylene Oxide Standard at 79,000 Da