Learn More
CELLTREAT Scientific Products Aspirating Pipette: 2 mL Capacity, Polystyrene, Individual Plastic/Plastic Wrapper Packed in Bags, Sterile, 400PK
SDP
Supplier:  CELLTREAT Scientific Products 229262A
Encompass_Preferred
Ideal for aspiration of media and sample waste removal without needing to measure the volume. Aspirating Pipets are supplied without graduations or plugs. Also available in 10, 25 and 50mL Individually Wrapped sizes. 2mL Aspirating Pipet available in plastic/plastic wrap (Part No. 229262A). Peel-to-open plastic/plastic wrappers are the ideal option for eliminating particulate; proven benefit over traditional paper/plastic wrapper.
Catalog No. NC1964624