Learn More
Medchemexpress LLC HY-15494 5mg Medchemexpress, Picropodophyllin CAS:477-47-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY154945MG
Encompass_Preferred
Medchemexpress, HY-15494 5mg Picropodophyllin CAS:477-47-4 Picropodophyllin (AXL1717) is a selective insulin-like growth factor-1 receptor (IGF-1R) inhibitor with an IC50 of 1 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9130
Due to product restrictions, please to purchase or view availability for this product.