Learn More
Perkin Elmer US LLC PerkinElmer ATR Heated Top Plate - Diamond, 60°C
Supplier: Perkin Elmer US LLC L1272380
Attenuated Total Reflectance (ATR) is a sampling technique which requires little or no sample preparation. A beam of infrared light passes throught the ATR crystal and is reflected off the surface of the sample. The reflection forms an evanescent wave which is collected and measured by the spectrometer. The ATR technique works for solid, liquid, and powder samples.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.