Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3B Scientific™ P93 PRO Male Catheterization Simulator
SDPSCI_ED

Procedures can be realistically demonstrated, practiced and assessed. 3B Scientific™ P93 PRO Male Catheterization Simulator has suprapubic catheter which students can practice along with the trainer.

Supplier:  3B Scientific™ P93PM

Catalog No. S37618


Add to Cart

Includes: Realistic lower abdominal model, Catheterization set with transparent bladder, Bottle and tube connectors and Carry case

Description

Description

  • Male catheterization with realistic resistance
  • Three levels of adjustable narrowing of urethra
  • Soft and movable foreskin
  • Anatomically realistic pelvic structure
  • Liquid outflows if catheterization is successfully carried out
  • Transparent bladder can be checked, and the abdominal wall can be removed
  • Includes suprapubic catheter for practicing, cleaning and caring
  • Stands securely on table with the help of non-slip feet
  • Magnetic connectors for quick set-up and dismantling
  • Easy to clean, maintain and can be completely taken apart
  • Sponge present inside prevents build-up of moisture

Warranty and Services

3 Year

Specifications

Specifications

PRO Male Simulator
50 x 42 x 30 cm
3.5 kg
Catheterization
7.71 lb.
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit