Learn More
BioMed Diagnostics A rapid 20-minute heat detection test to detect progesterone in cows milk. Allows you to know when the cow is in their estrous cycle. | Detects: Progesterone in cows milk. | Available in: 25 pack
Supplier:  BioMed Diagnostics 10000015
Encompass_Preferred
A rapid 20-minute heat detection test to detect progesterone in cows milk. The game-changing fast and accurate, near instant in-milk heat test used directly in your parlor. P4 Gold requires no special skills or training with its immediate visual reaction. Allows you to know when the cow is in their estrous cycle. | Detects: Progesterone in cows milk. | Available in: 25 pack
Catalog No. NC2582883