Learn More
Cole-Parmer Cole-Parmer P200-02 pH Meter Kit; 3-in-1 pH Electrode, Electrode Holder
Supplier:  Cole-Parmer 5925005
Encompass_Preferred
The P200 series of benchtop meters provides accurate, reliable results with the benefit of convenient accessories to make your lab run smoothly. Large color TFT display is easily visible, and features a stability icon to alert you when your readings are ready to be recorded and an electrode health icon indicating when you need to refill or replace your electrode.
Catalog No. NC1594405
Explore available promotions