Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dunlop™ Onguard™ Men's Flex 3 Work Boots

$133.00 - $143.00

Specifications

Certifications/Compliance ASTM F2413-05
Color Black
For Use With (Application) Used in oil rigs, refineries, food processing, general industrial areas, lawn care, dairies, breweries and utilities
Height (English) 16 in.
Lug Type Kick-off
View More Specs

Products 6
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-088-2087C
View Documents
Dunlop™
89908090
9
Pair for $143.00
Add to Cart
 
19-088-2087A
View Documents
Dunlop™
89908070
7
Pair for $143.00
Add to Cart
 
19-088-2087B
View Documents
Dunlop™
89908080
8
Pair for $143.00
Add to Cart
 
19-088-2087F
View Documents
Dunlop™
89908120
12
Pair for $143.00
Add to Cart
 
19-088-2087E
View Documents
Dunlop™
89908110
11
Pair for $133.00
Add to Cart
 
19-088-2087D
View Documents
Dunlop™
89908100
10
Pair for $143.00
Add to Cart
 
Description

Description

Specifications

Specifications

ASTM F2413-05
Used in oil rigs, refineries, food processing, general industrial areas, lawn care, dairies, breweries and utilities
Kick-off
Steel
Steel
Yes
Black
16 in.
PVC, Nitrile
Gray/yellow power-lug outsole and removable air cushion insole, slip-resistant
Fats, oils and chemicals
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit