Learn More
Immunology Consultants Normal Cat Serum, 100 ml
Supplier:  Immunology Consultants RS20100ML
Encompass_Preferred
Normal cat serum was collected from healthy cats and sterile filtered. Optimal working dilutions should be determined experimentally by the investigator.
Catalog No. 50-241-4291