Learn More
New England Biolabs, Inc. NEBNext Ultra™ II Directional RNA Second Strand Synthesis Module – 24 reactions
Supplier: New England Biolabs, Inc. E7550S
The NEBNext Ultra II Directional RNA Second Strand Synthesis Module has been optimized to generate double-stranded cDNA from first-strand cDNA, as part of the NEBNext Directional RNA library preparation workflow. This workflow uses the dUTP method for strand-specific library construction, which depends on incorporation of uracil into the second strand cDNA, enabled by the NEBNext Second Strand Synthesis Reaction Buffer with dUTP Mix, included in this module.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.