Learn More
Enzo Life Sciences Netrin-1 (human), (recombinant) (10µg)
Supplier: Enzo Life Sciences ALX522100C010
Concentration: 0.1mg/ml after reconstitution. Purity: ≥90% (SDS-PAGE). Formulation: Lyophilized. Contains PBS. Source: Produced in HEK 293 cells. The extracellular domain of human netrin-1 (aa 28-604) is fused at the N-terminus to a FLAG-tag. Specificity: Tests positive for binding to human Unc5b by ELISA. Previous lots produced from the same cell line bind to human, mouse and rat receptors UNC5H and DCC. Long Term Storage: -20°C. Applications: ELISA .
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.