Learn More
National Diagnostics™ Calci-Clear™ Rapid Decalcifying Solution
SDP
High-speed decalcifier that permits the processing of tissue samples in considerably less time
Supplier:  National Diagnostics™ HS1051GLL
Encompass
 View more versions of this product
Catalog No. 50-899-90141
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • High speed decalcification
  • Minimizes loss of tissue structure
  • Excellent staining characteristics
Specifications
Specifications
1 gal.
Product Suggestions
Product Suggestions