Learn More
Yamato Scientific America, Inc. Yamato 445L Natural and Forced Convection Glassware Drying Oven, with air in-take port filer, exhaust fan and germicidal lamp, 115V
Supplier: Yamato Scientific America, Inc. DG840C
445L glassware drying natural and forced convection oven with large observation window for easy observation. Equipped with filter at air in-take port, exhaust fan and germicidal lamp for fast drying. Highly efficient heat insulation material for both internal and external structure. Adjustable foot for stability on uneven floors. Equipped with stainless steel pipe heater and water receiving plate at the bottom. Stainless steel interior, easy to clean and highly resistant to corrosion. Dial setting and digital display of temperature control and timer. Can be used to store instruments after drying. Mobile on casters. Operating temp ambient +5~70°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.