Learn More
Yamato Scientific America, Inc. Yamato 445L Natural Convection Glassware Drying Oven, 115V
Supplier: Yamato Scientific America, Inc. DG800C
445L glassware drying natural convection oven with large observation window for easy observation. Highly efficient heat insulation material for both internal and external structure. Adjustable foot for stability on uneven floors. Equipped with stainless steel pipe heater and water receiving plate at the bottom. Stainless steel interior, easy to clean and highly resistant to corrosion. Dial setting and digital display of temperature control and timer. Can be used to store instruments after drying. Mobile on casters. Operating temp ambient +5~70°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.