Learn More
Yamato Scientific America, Inc. Yamato 153L Natural Convection Oven, 220V
Supplier: Yamato Scientific America, Inc. DX612C
Standard 153L natural convection constant temperature drying oven with extensive features and simple operation. Economical and cost saving. Easy to use and maintain. Excellent temperature accuracy. Digital PID controller, calibration off-set and increased safety and self diagnostic functions. Operating temp ambient +5~280°C