Learn More
Thermo Scientific™ Nalgene™ Syringe Filters, Non-sterile
Filter sample volumes of 10 to 100mL with these nonsterile syringe filters featuring a variety of membranes and pore sizes for most laboratory applications.
$266.00 - $3060.00
Catalog Number | Mfr. No. | Diameter (Metric) | Material (Membrane) | Pore Size | Sterility | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) | Material (Membrane) | Pore Size | Sterility | Price | Quantity & Availability | ||
09-740-101
|
Thermo Scientific™
7211320 |
13 mm | PTFE | 0.2 μm | Non-sterile |
Case of 100 for $450.00
|
|
||
09-740-102
|
Thermo Scientific™
7211345 |
13 mm | PTFE | 0.45 μm | Non-sterile |
Case of 100 for $450.00
|
|
||
09-740-103
|
Thermo Scientific™
7222000 |
25 mm | Glass Fiber | 1.1 μm | Non-sterile |
Case of 50 for $266.00
|
|
||
09-740-110
|
Thermo Scientific™
7242020 |
25 mm | SFCA | 0.2 μm | Non-sterile |
Case of 500
for $2,043.65
Save 22% Reg : |
|
||
09-740-111
|
Thermo Scientific™
7242045 |
25 mm | SFCA | 0.45 μm | Non-sterile |
Case of 500 for $2,620.00
|
|
||
09-740-112
|
Thermo Scientific™
7242080 |
25 mm | Cellulose Acetate | 0.8 μm | Non-sterile |
Case of 500 for $2,140.00
|
|
||
09-740-117
|
Thermo Scientific™
7272020 |
25 mm | Nylon | 0.2 μm | Non-sterile |
Case of 500 for $2,400.00
|
|
||
09-740-118
|
Thermo Scientific™
7272045 |
25 mm | Nylon | 0.45 μm | Non-sterile |
Case of 500
for $2,018.65
Save 23% Reg : |
|
||
09-740-119
|
Thermo Scientific™
7282020 |
25 mm | PTFE | 0.2 μm | Non-sterile |
Case of 500 for $3,060.00
|
|
||
09-740-120
|
Thermo Scientific™
7282045 |
25 mm | PTFE | 0.45 μm | Non-sterile |
Case of 500
for $2,380.65
Save 22% Reg : |
|
||
09-740-34C
|
Thermo Scientific™
1760020 |
4 mm | Nylon | 0.2 μm | Non-sterile |
Pack of 100 for $623.50
Case of 4 PK for $1,380.00
|
|
||
09-740-34D
|
Thermo Scientific™
1760045 |
4 mm | Nylon | 0.45 μm | Non-sterile |
Pack of 100
for $541.65
Save 15% Reg :
Case of 4 PK
for $1,183.65
Save 15% Reg : |
|
||
Description
Thermo Scientific™ Nalgene™ syringe filters are built to meet your lab’s clarification and sterilization needs. Our filters are available in a variety of sizes and membranes that can be easily used for both sterile and non-sterile laboratory applications. Whether your research necessitates prefiltration, sample preparation, sterile filtration, laboratory filtration, or gas venting, Nalgene syringe filters are lab essentials.
Key Features- 10 to 100mL sample size
- Inlet: Female Luer-Lok™
- Outlet: Male Luer slip
- Polypropylene housing with ID color rings
- Pore size and membrane type printed on unit
Surfactant-free Cellulose Acetate (SFCA)
- Low protein binding, lower extractables than standard CA
- Good for cell culture fluids and cellular solutions
- Polypropylene housings with blue color rings
Cellulose Acetate (CA)
- Some protein binding and extractables
- For sterile filtration and clarification of: buffers, aqueous solutions and reagents
- Polypropylene housings with red color rings
Nylon
- Naturally hydrophilic, low extractables, some protein binding
- Useful for alcohols, cryoprotectants such as DMSO and with aqueous/biological and weak solvents
- Polypropylene housings with pink color rings
- Naturally hydrophobic
- Excellent for HPLC/GC solvent filtration. Ideal for sterile venting of air or other gases during filtration
- Polypropylene housings with white color rings
Specifications
Non-sterile | |
Syringe Filter |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.