Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
N-Cinnamoylglycine, ≥98% (HPLC), MilliporeSigma™ Supelco™
Supplier: MilliporeSigma Supelco 9895610MG
Specifications
16534-24-0 | |
Analytical Standard | |
MFCD00136326 | |
205.21 | |
Analytical Standard |
10 mg | |
C11H11 NO3 | |
N-(1-Oxo-3-phenyl-2-propen-1-yl)glycine; Cinnamoyl glycine | |
≥98% (HPLC) |
Safety and Handling
ShelfLife : Limited shelf life, expiry date on the label
Recommended Storage : 2°C to 8°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction