Learn More
US Biomax Inc MULTI-NORMAL ORGANS TISSUE ARR
SDP
Supplier:  US Biomax Inc FDA331
Encompass
Multi-normal organs tissue array,representing FDA guideline for antibody cross-reactivity testing,32 cases/33 cores(core size 1.5mm)
Catalog No. NC2253321