Learn More
Multi-Element Organometallic Oil Standards 23 Element, 100μg/g; SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
$250.00 - $387.00
Specifications
Chemical Name or Material | 23 Element Oil Standard |
---|---|
Packaging | Triple-leached LDPE bottle |
Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
Analyte or Component Names | Al,Ba,B |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
S23100Z
|
SPEX CertiPrep
S23100Z |
50 g | Triple-leached LDPE bottle |
Each for $250.00
|
|
||||
S23100Y
|
SPEX CertiPrep
S23100Y |
100 g | Triple-leached LDPE bottle |
Each for $387.00
|
|
||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Ag | 100μg/g |
Al | 100μg/g |
B | 100μg/g |
Ba | 100μg/g |
Ca | 100μg/g |
Cd | 100μg/g |
Cr | 100μg/g |
Cu | 100μg/g |
Fe | 100μg/g |
K | 100μg/g |
Mg | 100μg/g |
Mn | 100μg/g |
Mo | 100μg/g |
Na | 100μg/g |
Ni | 100μg/g |
P | 100μg/g |
Pb | 100μg/g |
Sb | 100μg/g |
Si | 100μg/g |
Sn | 100μg/g |
Ti | 100μg/g |
V | 100μg/g |
Zn | 100μg/g |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
23 Element Oil Standard | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
Al,Ba,B |
Triple-leached LDPE bottle | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
Base Oil 75 |
Safety and Handling
Causes skin irritation, serious eye irritation
DOTInformation : Nonhazardous
Recommended Storage : Ambient
Product intended for laboratory use only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.