Learn More
Tocris Bioscience™ MRS 2578
Selective P2Y6 antagonist
$182.12 - $755.16
Specifications
Product Type | MRS 2578 |
---|---|
Molecular Weight (g/mol) | 472.66 |
Inhibitors | MRS 2578 |
Description
Selective antagonist of P2Y6 nucleotide receptors; IC50 values are 37 and 98 nM at human and rat P2Y6 receptors respectively. Displays no activity at P2Y1, P2Y2, P2Y4 and P2Y11 receptors (IC50 > 10 μM). Inhibits agonist-induced cardiomyocyte contraction and UDP-induced phagocytosis.Specifications
MRS 2578 | |
472.66 | |
MRS 2578 |