Learn More
MilliporeSigma Mouse Serum, 100 mL
Supplier: MilliporeSigma S25100ML
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Non-sterile mouse serum collected from normal healthy mice. Used for blocking of interfering antibodies.