Learn More
PBL Assay Science Mouse Interferon Alpha 4, mammalian (IFN-Alpha 4) [1e5 (100,000) units/vial]
Supplier: PBL Assay Science 121151
Recombinant protein Mouse Interferon Alpha 4, mammalian is manufactured by PBL Assay Science (Catalog #12115-1) for research use only (RUO) and packaged in 1e5 (100,000) units/vial. Mu IFN-Alpha 4 exhibits earlier endogenous expression than other Mu Type I IFNs.