Learn More
Enzo Life Sciences Matrix Metalloproteinase-8 (MMP-8) colorimetric drug discovery kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences BMLAK4140001
Encompass_Preferred
A QUANTIZYME Assay System. The MMP-8 Colorimetric Drug Discovery Kit is a complete assay system designed to screen inhibitors of matrix metalloproteinase-8 (MMP-8, neutrophil collagenase, collagenase-2) using a thiopeptide as a chromogenic substrate. The assays are performed in a convenient 96-well plate format. Also included are active enzyme, a prototypic control inhibitor (NNGH), and a detailed instruction booklet. Alternative name: Neutrophil collagenase, Collagenase 2. Applications: Colorimetric detection, HTS Activity assay.
Catalog No. 50-201-0274