Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Supertek™ Microscope Slide Boxes
Supertek™ Microscope Slide Boxes are designed for the safe long term storage of microscope slides. These storage boxes accommodate standard 25 x 75 mm /1 x 3 in. microscope slides.
$4.90 - $23.75
Catalog Number | Mfr. No. | Slide Capacity | Dimensions (L x W x H) | Color | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Slide Capacity | Dimensions (L x W x H) | Color | Price | Quantity & Availability | |||
S160116
|
Supertek™
CH11191/11 |
12 | 3.25 x 2 x 1.25 in. | White |
Each for $4.90
|
|
|||
S160117
|
Supertek™
CH11191/12 |
25 | 3.25 x 4 x 1.25 in. | Blue |
Each for $7.90
|
|
|||
S160118
|
Supertek™
CH11191/13 |
50 | 3.25 x 8 x 1.25 in. | Gray |
Each for $12.75
|
|
|||
S160119
|
Supertek™
CH11191/14 |
100 | 7 x 8 x 1.25 in. | Black |
Each for $23.75
|
|
|||
Description
- Made of durable high quality ABS plastic.
- Heavy walls will not wrap, splinter or crack.
- Cork sheet lined base for safe storage.
- Boxes are unaffected by humidity & also are insect proof.
- Available in 4 different capacities for storage of 12/25/50/100 slides.
- An inventory sheet is located on the inside cover for convenient slide identification and organization.
Specifications
Microscope slides |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title