Learn More
Kamiya Biomedical Microalbumin Reagent, 80mL+20mL/PK RX
Supplier: Kamiya Biomedical KAI019
K-ASSAY Microalbumin Assay Reagent. R1: 4 x 20mL (liquid), R2: 2 x 10mL (liquid). Assay Range: 0.2 - 30.0 mg/dL. The K-ASSAY Microalbumin assay is intended for the quantitative determination of human albumin in urine by immunoturbidimetric assay. Measurement of albumin in urine aids in the diagnosis of kidney dysfunction. FOR IN VITRO DIAGNOSTIC USE. Required Calibrator: K-ASSAY Microalbumin Calibrator (Cat. No. KAI-020C). Recommended Controls: K-ASSAY Microalbumin Urine Control (Cat. No. K37C-4M). This assay is intended for use on most popular chemistry analyzers that allow open channel reagents. Please contact KAMIYA BIOMEDICAL COMPANY to check parameter availability for your analyzer.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.