Learn More
Cole-Parmer Extech 407860 Heavy Duty Vibration/Acceleration/Velocity/Displacement Meter
Supplier: Cole-Parmer 9883130
Connect the vibration sensor to equipment via magnetic attachment or threaded stud mount. The RECORD feature stores min and max measurements for later recall, Hold feature freezes display, and auto-off function saves battery life. Download stored measurements to your PC via RS-232 interface (sold separately).