Learn More
Microsolv Technology Corporation Metabolomics Development Kit, 2.o, 2x150. 1 Cogent Diamond Hydride 2.o™ & 1 Cogent Bidentate C18 2.o™. 1 PK.
Supplier:  Microsolv Technology Corporation 4320015P2
Encompass_Preferred
This is an economical way to purchase two HPLC Columns that are often used with LCMS in Metabolomics and many other fields. Each Column has a 2.1mm Internal Diameter and is 150mm Long, packed with 2.2um particles and 120A pore size and are manufactured with Cogent TYPE-C™ Silica Hydride. Includes the Cogent Diamond Hydride™ and Cogent Bidentate C18™ columns.
Catalog No. 50-227-0856