Learn More
Microsolv Technology Corporation Metabolomics Development Kit, 2.o, 2.1x50. 1 Cogent Diamond Hydride 2.o™ & 1 Cogent Bidentate C18 2.o™ 2.1mm 50mm. 1 PK.
Supplier: Microsolv Technology Corporation 4320005P2
This is an economical way to purchase two HPLC Columns that are often used with LCMS in Metabolomics and many other fields. Each Column has a 2.1mm Internal Diameter and is 50mm Long, packed with 2.2um particles and 120A pore size and are manufactured with Cogent TYPE-C™ Silica Hydride. Includes the Cogent Diamond Hydride™ and Cogent Bidentate C18™ columns.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.