Learn More
3M™ Mechanical Mount HF RFID Tag with Cable Ties
3M™ Mechanical Mount HF RFID Tag with Cable Ties is designed to be mechanically mounted to surfaces
Supplier: 3M™ 7100107839
Description
- Durable, heavy-duty construction
- Utilizes passive high frequency 13.56 MHz technology and is pre-programmed unique ID
- Ready for use with 3M™ Inspection and Asset Management System
- Best suited for metal surfaces.
- 3M Catalog# RFID-MMTB
Specifications
50/Pk. | |
Black | |
Connected Safety |
RFID Tag | |
Polyamide with Glass Fiber | |
RFID-MMTB |