Learn More
Creative Biomart Recombinant Lassa GP1 Antigen, His-tagged
Supplier: Creative Biomart MSP1336
The E. coli derived Recombinant Lassa Glycoprotein-1 (strain Mouse/Sierra Leone/Josiah/1976) containing 205 amino acids, having an Mw of 23 kDa and the Isoelectric point is 6.7. The Lassa GP1 protein