Learn More
Medchemexpress LLC HY-13672 5mg Medchemexpress, LY2334737 CAS:892128-60-8 Purity:>98%
Supplier: Medchemexpress LLC HY136725MG
Medchemexpress, HY-13672 5mg LY2334737 CAS:892128-60-8 LY2334737 is an nucleoside analog and is an orally active prodrug of Gemcitabine. LY2334737 exhibits inhibitory activity against enterovirus A71 (EV-A71) infection. LY2334737 has antiviral and anticancer effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.