Learn More
Chemglass Life Sciences Vessel, Bio Reactor, 6L, Low Profile, Graduated, 100-440 GPI Thread CN,(4) Vertical GL-45 SN's
Supplier: Chemglass Life Sciences CLS140406
Vessel, Bio Reactor, 6L, Low Profile, Graduated, 100-440 GPI Thread CN,(4) Vertical GL-45 SN's. Vessel has four vertical GL-45 threaded side necks and a 100mm threaded center neck. The 3 and 6 liter vessels are low profile to maximize incubator space. Supplied complete with (4) solid GL-45 caps and (4) pouring rings.