Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ L-755,507
Very potent and selective β3 partial agonist
$272.60 - $1141.44
Specifications
Purity | >98% |
---|---|
Product Type | L-755,507 |
Molecular Weight (g/mol) | 584.73 |
Inhibitors | L-755,507 |
Description
Potent β3-adrenergic receptor partial agonist > 1000-fold selective over β1- and β2-adrenoceptors (EC50 values are 0.43, 580 and > 10000 nM for activation of cloned human β3-, β1- and β2-adrenoceptors respectively). Stimulates lipolysis in rhesus adipocytes in vitro (EC50 = 3.9 nM). Enhances CRISPR-mediated homology-directed repair (HDR) efficiency 2-3-fold for large fragments and ∽9-fold for point mutations, in human induced pluripotent stem cells (iPSCs).Specifications
>98% | |
L-755,507 | |
584.73 | |
L-755,507 |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title